Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 7 - 8 923 68
NM_001313727.1 NP_001300656.1 3 - 4 481 68
NM_031418.3 NP_113606.2 6 - 7 831 68
XM_017018118.1 XP_016873607.1 4 - 5 459 68
GenBank mRNA AJ300461.1 - 6 - 7 1538 68
AK295143.1 - 3 - 4 394 68
AK295816.1 - 3 - 4 393 68
AK304781.1 - 7 - 8 764 68
AK316018.1 - 3 - 4 393 68
BP219022.1 - 3 - 4 482 68

View Full Product Details