Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 9 - 10 1105 81
NM_001313727.1 NP_001300656.1 5 - 6 663 81
NM_031418.3 NP_113606.2 8 - 9 1013 81
XM_017018118.1 XP_016873607.1 6 - 7 641 81
XM_017018119.1 XP_016873608.1 3 - 4 253 81
GenBank mRNA AJ300461.1 - 8 - 9 1720 81
AK295143.1 - 5 - 6 576 81
AK295816.1 - 5 - 6 575 81
AK302659.1 - 8 - 9 1052 81
AK304781.1 - 9 - 10 946 81
AK316018.1 - 5 - 6 575 81

View Full Product Details