The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029881.2 | NP_001025052.1 | 5 - 6 | 494 | 85 |
XM_011532514.2 | XP_011530816.1 | 5 - 6 | 577 | 85 | |
XM_011532517.2 | XP_011530819.1 | 3 - 4 | 864 | 85 | |
XM_017003329.1 | XP_016858818.1 | 4 - 5 | 1348 | 85 | |
XM_017003330.1 | XP_016858819.1 | 3 - 4 | 1258 | 85 | |
GenBank mRNA | AW138422.1 | - | 2 - 3 | 85 | |
BC127684.1 | - | 5 - 6 | 494 | 85 | |
BC127685.1 | - | 5 - 6 | 492 | 85 | |
BC140923.1 | - | 5 - 6 | 468 | 85 |