Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH15
Entrez Gene ID
65217
Gene Name
protocadherin related 15
Gene Aliases
CDHR15, DFNB23, USH1F
Chromosome Location
Chr.10: 53802771 - 55629182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142763.1 NP_001136235.1 15 - 16 2192 95
NM_001142764.1 NP_001136236.1 14 - 15 2177 95
NM_001142766.1 NP_001136238.1 14 - 15 2177 95
NM_001142767.1 NP_001136239.1 13 - 14 2066 95
NM_001142768.1 NP_001136240.1 13 - 14 2111 95
NM_001142769.1 NP_001136241.1 16 - 17 2213 95
NM_001142770.1 NP_001136242.1 14 - 15 2177 95
NM_001142771.1 NP_001136243.1 15 - 16 2192 95
NM_001142772.1 NP_001136244.1 14 - 15 2177 95
NM_001142773.1 NP_001136245.1 13 - 14 2111 95
NM_033056.3 NP_149045.3 14 - 15 2177 95
XM_017016571.1 XP_016872060.1 14 - 15 2775 95
XM_017016572.1 XP_016872061.1 14 - 15 2773 95
XM_017016573.1 XP_016872062.1 21 - 22 5574 95
GenBank mRNA AK126923.1 - 16 - 17 2078 95
AL834134.1 - 14 - 15 2074 95
AY029205.1 - 14 - 15 2165 95
AY029237.1 - 14 - 15 2177 95
EU718480.1 - 15 - 16 2198 95
EU718481.1 - 14 - 15 2177 95
EU718482.1 - 14 - 15 2177 95

View Full Product Details