Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH15
Entrez Gene ID
65217
Gene Name
protocadherin related 15
Gene Aliases
CDHR15, DFNB23, USH1F
Chromosome Location
Chr.10: 53802771 - 55629182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142763.1 NP_001136235.1 6 - 7 885 98
NM_001142764.1 NP_001136236.1 5 - 6 870 98
NM_001142765.1 NP_001136237.1 5 - 6 870 98
NM_001142766.1 NP_001136238.1 5 - 6 870 98
NM_001142767.1 NP_001136239.1 5 - 6 870 98
NM_001142768.1 NP_001136240.1 4 - 5 804 98
NM_001142769.1 NP_001136241.1 6 - 7 885 98
NM_001142770.1 NP_001136242.1 5 - 6 870 98
NM_001142771.1 NP_001136243.1 6 - 7 885 98
NM_001142772.1 NP_001136244.1 5 - 6 870 98
NM_001142773.1 NP_001136245.1 4 - 5 804 98
NM_033056.3 NP_149045.3 5 - 6 870 98
XM_017016571.1 XP_016872060.1 5 - 6 1468 98
XM_017016572.1 XP_016872061.1 5 - 6 1466 98
XM_017016573.1 XP_016872062.1 12 - 13 4267 98
GenBank mRNA AK126923.1 - 7 - 8 771 98
AL834134.1 - 5 - 6 767 98
AY029205.1 - 5 - 6 858 98
AY029237.1 - 5 - 6 870 98
EU718480.1 - 5 - 6 870 98
EU718481.1 - 5 - 6 870 98
EU718482.1 - 5 - 6 870 98

View Full Product Details