Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH15
Entrez Gene ID
65217
Gene Name
protocadherin related 15
Gene Aliases
CDHR15, DFNB23, USH1F
Chromosome Location
Chr.10: 53802771 - 55629182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142763.1 NP_001136235.1 11 - 12 1508 81
NM_001142764.1 NP_001136236.1 10 - 11 1493 81
NM_001142765.1 NP_001136237.1 10 - 11 1493 81
NM_001142766.1 NP_001136238.1 10 - 11 1493 81
NM_001142767.1 NP_001136239.1 9 - 10 1382 81
NM_001142768.1 NP_001136240.1 9 - 10 1427 81
NM_001142769.1 NP_001136241.1 11 - 12 1508 81
NM_001142770.1 NP_001136242.1 10 - 11 1493 81
NM_001142771.1 NP_001136243.1 11 - 12 1508 81
NM_001142772.1 NP_001136244.1 10 - 11 1493 81
NM_001142773.1 NP_001136245.1 9 - 10 1427 81
NM_033056.3 NP_149045.3 10 - 11 1493 81
XM_017016571.1 XP_016872060.1 10 - 11 2091 81
XM_017016572.1 XP_016872061.1 10 - 11 2089 81
XM_017016573.1 XP_016872062.1 17 - 18 4890 81
GenBank mRNA AK126923.1 - 12 - 13 1394 81
AL834134.1 - 10 - 11 1390 81
AY029205.1 - 10 - 11 1481 81
AY029237.1 - 10 - 11 1493 81
EU718480.1 - 10 - 11 1493 81
EU718481.1 - 10 - 11 1493 81
EU718482.1 - 10 - 11 1493 81

View Full Product Details