The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015316.2 | NP_056131.2 | 1 - 2 | 291 | 105 |
XM_017021116.1 | XP_016876605.1 | 1 - 2 | 332 | 105 | |
GenBank mRNA | AJ318887.2 | - | 1 - 2 | 166 | 105 |
BC035446.1 | - | 1 - 2 | 260 | 105 | |
BC136527.1 | - | 1 - 2 | 95 | 105 | |
DB461586.2 | - | 1 - 2 | 291 | 105 |