The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015316.2 | NP_056131.2 | 7 - 8 | 1106 | 119 |
XM_005267487.4 | XP_005267544.1 | 7 - 8 | 1510 | 119 | |
XM_011536593.2 | XP_011534895.1 | 7 - 8 | 1510 | 119 | |
XM_017021116.1 | XP_016876605.1 | 7 - 8 | 1147 | 119 | |
XM_017021117.1 | XP_016876606.1 | 7 - 8 | 923 | 119 | |
XM_017021118.1 | XP_016876607.1 | 7 - 8 | 836 | 119 | |
GenBank mRNA | AB018314.1 | - | 3 - 4 | 397 | 119 |
AJ318887.2 | - | 7 - 8 | 981 | 119 | |
AK125913.1 | - | 8 - 9 | 1542 | 119 | |
BC035446.1 | - | 7 - 8 | 1075 | 119 | |
BC136527.1 | - | 7 - 8 | 910 | 119 |