The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152474.4 | NP_689687.1 | 1 - 2 | 223 | 78 |
GenBank mRNA | BC033933.2 | - | 1 - 2 | 226 | 78 |