Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MALT1
Entrez Gene ID
10892
Gene Name
MALT1 paracaspase
Gene Aliases
IMD12, MLT, MLT1, PCASP1
Chromosome Location
Chr.18: 58671386 - 58753806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006785.3 NP_006776.1 13 - 14 1860 157
NM_173844.2 NP_776216.1 12 - 13 1827 157
GenBank mRNA AB026118.1 - 12 - 13 1634 157
AF130356.2 - 13 - 14 1766 157
AF316597.1 - 13 - 14 1860 157
AK092004.1 - 2 - 3 1184 157
AK291386.1 - 13 - 14 1826 157
BC030143.2 - 12 - 13 1681 157

View Full Product Details