The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006785.3 | NP_006776.1 | 7 - 8 | 1212 | 126 |
XM_011525794.1 | XP_011524096.1 | 7 - 8 | 1178 | 126 | |
GenBank mRNA | AF130356.2 | - | 7 - 8 | 1118 | 126 |
AF316597.1 | - | 7 - 8 | 1212 | 126 | |
AK291386.1 | - | 7 - 8 | 1178 | 126 |