The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145026.3 | NP_659463.1 | 4 - 5 | 515 | 103 |
GenBank mRNA | AK058171.1 | - | 3 - 4 | 761 | 103 |
BC100956.2 | - | 3 - 4 | 570 | 103 | |
BC100957.2 | - | 3 - 4 | 570 | 103 | |
BC100958.2 | - | 3 - 4 | 570 | 103 | |
BC100959.2 | - | 4 - 5 | 664 | 103 | |
BC110065.1 | - | 3 - 4 | 570 | 103 | |
BC111414.1 | - | 3 - 4 | 570 | 103 |