The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145026.3 | NP_659463.1 | 5 - 6 | 679 | 71 |
GenBank mRNA | AK058171.1 | - | 4 - 5 | 925 | 71 |
BC100956.2 | - | 4 - 5 | 734 | 71 | |
BC100957.2 | - | 4 - 5 | 734 | 71 | |
BC100958.2 | - | 4 - 5 | 734 | 71 | |
BC100959.2 | - | 5 - 6 | 828 | 71 | |
BC110065.1 | - | 4 - 5 | 734 | 71 | |
BC111414.1 | - | 4 - 5 | 734 | 71 |