The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003945.1 | 1 - 1 | 7324 | 173 | |
GenBank mRNA | AK023435.1 | - | 1 - 1 | 1909 | 173 |
AL834507.1 | - | 1 - 1 | 1975 | 173 | |
BX538342.1 | - | 1 - 1 | 3426 | 173 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AU155698.1 | - | 1 | Chr. 11 - 6714340 - 6714799 [+] on Build GRCh38 | 173 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial ribosomal protein L17 | MRPL17 | L17mt, LIP2, MRP-L17, MRP-L26, RPL17L, RPML26 | -29.7kb | Sense | Chr. - 6680385 - 6683401 on Build GRCh38 |
Downstream | olfactory receptor family 2 subfamily AG member 2 | OR2AG2 | OR11-76, OR2AG2P | 46.1kb | Sense | Chr. - 6768007 - 6768957 on Build GRCh38 |