Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGLYRP4
Entrez Gene ID
57115
Gene Name
peptidoglycan recognition protein 4
Gene Aliases
PGLYRPIbeta, PGRP-Ibeta, PGRPIB, SBBI67
Chromosome Location
Chr.1: 153327410 - 153348844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020393.3 NP_065126.2 4 - 5 712 79
XM_011509789.2 XP_011508091.1 4 - 5 724 79
XM_011509790.1 XP_011508092.1 4 - 5 712 79
XM_011509791.2 XP_011508093.1 4 - 5 703 79
XM_011509792.1 XP_011508094.1 4 - 5 712 79
GenBank mRNA AF242518.1 - 4 - 5 428 79
AK292203.1 - 4 - 5 715 79
AY035377.1 - 4 - 5 417 79
BC107157.1 - 3 - 4 399 79
BC107158.1 - 3 - 4 399 79
BC142636.1 - 4 - 5 687 79

View Full Product Details