The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301110.1 | NP_001288039.1 | 16 - 17 | 2213 | 79 |
NM_207517.2 | NP_997400.2 | 16 - 17 | 2213 | 79 | |
XM_011521822.2 | XP_011520124.1 | 17 - 18 | 2285 | 79 | |
XM_011521823.2 | XP_011520125.1 | 17 - 18 | 2285 | 79 | |
XM_011521824.2 | XP_011520126.1 | 17 - 18 | 2285 | 79 | |
XM_011521825.2 | XP_011520127.1 | 17 - 18 | 2285 | 79 | |
XM_017022434.1 | XP_016877923.1 | 17 - 18 | 2285 | 79 | |
XM_017022435.1 | XP_016877924.1 | 16 - 17 | 2081 | 79 | |
GenBank mRNA | AF237652.1 | - | 16 - 17 | 79 | |
AK308845.1 | - | 16 - 17 | 79 | ||
BC128389.1 | - | 16 - 17 | 79 | ||
BC128390.1 | - | 16 - 17 | 79 |