The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300752.1 | NP_001287681.1 | 5 - 6 | 1395 | 111 |
NM_001300759.1 | NP_001287688.1 | 6 - 7 | 1414 | 111 | |
NM_018700.3 | NP_061170.2 | 6 - 7 | 1630 | 111 | |
XM_017009621.1 | XP_016865110.1 | 8 - 9 | 1642 | 111 | |
XM_017009622.1 | XP_016865111.1 | 8 - 9 | 2359 | 111 | |
XM_017009623.1 | XP_016865112.1 | 8 - 9 | 1642 | 111 | |
GenBank mRNA | AB196558.1 | - | 5 - 6 | 886 | 111 |
AJ272269.1 | - | 6 - 7 | 1247 | 111 | |
AK058179.1 | - | 1 - 2 | 271 | 111 | |
AK296389.1 | - | 6 - 7 | 1412 | 111 | |
AK316422.1 | - | 6 - 7 | 1317 | 111 | |
BC130334.1 | - | 6 - 7 | 1299 | 111 |