Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
Chromosome Location
Chr.5: 41925249 - 42040230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297437.1 NP_001284366.1 1 - 2 243 100
NM_012176.2 NP_036308.1 1 - 2 241 100
NM_033484.2 NP_277019.1 1 - 2 241 100
XM_011514026.2 XP_011512328.1 1 - 2 246 100
GenBank mRNA AF176703.1 - 1 - 2 179 100
AK024075.1 - 1 - 2 241 100
AU143782.1 - 1 - 2 241 100
BC040011.1 - 1 - 2 202 100
BC048098.1 - 1 - 2 209 100

View Full Product Details