The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284301.2 | NP_001271230.1 | 15 - 16 | 2764 | 117 |
NM_001284302.2 | NP_001271231.1 | 15 - 16 | 2761 | 117 | |
NM_018685.4 | NP_061155.2 | 16 - 17 | 2875 | 117 | |
XM_006715746.1 | XP_006715809.1 | 16 - 17 | 2872 | 117 | |
XM_006715747.3 | XP_006715810.1 | 15 - 16 | 2761 | 117 | |
XM_017012354.1 | XP_016867843.1 | 16 - 17 | 2860 | 117 | |
XM_017012355.1 | XP_016867844.1 | 16 - 17 | 2860 | 117 | |
XM_017012356.1 | XP_016867845.1 | 15 - 16 | 2749 | 117 | |
GenBank mRNA | AF273437.1 | - | 16 - 17 | 2810 | 117 |
AK001468.1 | - | 6 - 7 | 794 | 117 | |
AK001472.1 | - | 11 - 12 | 1542 | 117 | |
AK023208.1 | - | 7 - 8 | 916 | 117 | |
AK291254.1 | - | 15 - 16 | 2693 | 117 | |
AK299803.1 | - | 16 - 17 | 2577 | 117 | |
BC034692.1 | - | 5 - 6 | 571 | 117 | |
BC070066.1 | - | 15 - 16 | 2746 | 117 | |
CR936650.1 | - | 13 - 14 | 1965 | 117 |