The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284301.2 | NP_001271230.1 | 7 - 8 | 1620 | 150 |
NM_001284302.2 | NP_001271231.1 | 7 - 8 | 1620 | 150 | |
NM_018685.4 | NP_061155.2 | 7 - 8 | 1620 | 150 | |
XM_006715746.1 | XP_006715809.1 | 7 - 8 | 1617 | 150 | |
XM_006715747.3 | XP_006715810.1 | 7 - 8 | 1617 | 150 | |
XM_017012354.1 | XP_016867843.1 | 7 - 8 | 1617 | 150 | |
XM_017012355.1 | XP_016867844.1 | 7 - 8 | 1617 | 150 | |
XM_017012356.1 | XP_016867845.1 | 7 - 8 | 1617 | 150 | |
GenBank mRNA | AF273437.1 | - | 7 - 8 | 1555 | 150 |
AK001472.1 | - | 3 - 4 | 398 | 150 | |
AK291254.1 | - | 7 - 8 | 1552 | 150 | |
AK299803.1 | - | 7 - 8 | 1322 | 150 | |
BC070066.1 | - | 7 - 8 | 1602 | 150 | |
CR936650.1 | - | 4 - 5 | 710 | 150 |