The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256721.1 | NP_001243650.1 | 10 - 11 | 1333 | 122 |
NM_006430.3 | NP_006421.2 | 11 - 12 | 1423 | 122 | |
GenBank mRNA | AF026291.1 | - | 11 - 12 | 1361 | 122 |
AK129860.1 | - | 11 - 12 | 1332 | 122 | |
AK290538.1 | - | 11 - 12 | 1390 | 122 | |
AK294665.1 | - | 10 - 11 | 1167 | 122 | |
AK295330.1 | - | 11 - 12 | 1196 | 122 | |
AK303082.1 | - | 10 - 11 | 1318 | 122 | |
AK307856.1 | - | 12 - 13 | 1357 | 122 | |
AK308942.1 | - | 11 - 12 | 1340 | 122 | |
AK312586.1 | - | 11 - 12 | 1365 | 122 | |
AK315975.1 | - | 12 - 13 | 1599 | 122 | |
BC014676.1 | - | 4 - 5 | 393 | 122 | |
BC106933.1 | - | 11 - 12 | 1356 | 122 | |
BC106934.1 | - | 11 - 12 | 1359 | 122 | |
U38846.1 | - | 11 - 12 | 1259 | 122 |