The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020393.3 | NP_065126.2 | 3 - 4 | 506 | 72 |
XM_011509789.2 | XP_011508091.1 | 3 - 4 | 518 | 72 | |
XM_011509790.1 | XP_011508092.1 | 3 - 4 | 506 | 72 | |
XM_011509792.1 | XP_011508094.1 | 3 - 4 | 506 | 72 | |
GenBank mRNA | AK292203.1 | - | 3 - 4 | 509 | 72 |
AY035377.1 | - | 3 - 4 | 211 | 72 | |
BC107157.1 | - | 2 - 3 | 193 | 72 | |
BC107158.1 | - | 2 - 3 | 193 | 72 | |
BC142636.1 | - | 3 - 4 | 481 | 72 |