Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCCA
Entrez Gene ID
5095
Gene Name
propionyl-CoA carboxylase alpha subunit
Gene Aliases
-
Chromosome Location
Chr.13: 100089015 - 100530437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000282.3 NP_000273.2 6 - 7 578 151
NM_001127692.2 NP_001121164.1 5 - 6 500 151
NM_001178004.1 NP_001171475.1 6 - 7 578 151
XM_005254059.2 XP_005254116.1 6 - 7 573 151
XM_011521093.1 XP_011519395.1 6 - 7 573 151
XM_017020605.1 XP_016876094.1 6 - 7 573 151
XM_017020606.1 XP_016876095.1 5 - 6 424 151
XM_017020607.1 XP_016876096.1 6 - 7 389 151
XM_017020608.1 XP_016876097.1 6 - 7 573 151
XM_017020609.1 XP_016876098.1 6 - 7 486 151
XM_017020610.1 XP_016876099.1 4 - 5 840 151
XM_017020611.1 XP_016876100.1 6 - 7 573 151
XM_017020612.1 XP_016876101.1 6 - 7 573 151
XM_017020613.1 XP_016876102.1 6 - 7 573 151
XM_017020614.1 XP_016876103.1 5 - 6 424 151
XM_017020615.1 XP_016876104.1 6 - 7 573 151
XM_017020616.1 XP_016876105.1 6 - 7 573 151
XM_017020617.1 XP_016876106.1 3 - 4 236 151
XM_017020618.1 XP_016876107.1 3 - 4 236 151
GenBank mRNA AF385926.1 - 6 - 7 510 151
AK225288.1 - 6 - 7 478 151
AK225299.1 - 6 - 7 476 151
AK296771.1 - 5 - 6 398 151
AK298318.1 - 6 - 7 500 151
AK315503.1 - 6 - 7 476 151
BC000140.2 - 6 - 7 450 151
DA781564.1 - 5 - 6 398 151
X14608.1 - 6 - 7 445 151
Z36837.1 - 2 - 3 172 151

View Full Product Details