Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCCA
Entrez Gene ID
5095
Gene Name
propionyl-CoA carboxylase alpha subunit
Gene Aliases
-
Chromosome Location
Chr.13: 100089015 - 100530437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000282.3 NP_000273.2 8 - 9 743 64
NM_001127692.2 NP_001121164.1 7 - 8 665 64
NM_001178004.1 NP_001171475.1 8 - 9 743 64
XM_005254059.2 XP_005254116.1 8 - 9 738 64
XM_011521093.1 XP_011519395.1 8 - 9 738 64
XM_017020605.1 XP_016876094.1 8 - 9 738 64
XM_017020606.1 XP_016876095.1 7 - 8 589 64
XM_017020607.1 XP_016876096.1 8 - 9 554 64
XM_017020608.1 XP_016876097.1 8 - 9 738 64
XM_017020609.1 XP_016876098.1 8 - 9 651 64
XM_017020610.1 XP_016876099.1 6 - 7 1005 64
XM_017020611.1 XP_016876100.1 8 - 9 738 64
XM_017020612.1 XP_016876101.1 8 - 9 738 64
XM_017020613.1 XP_016876102.1 8 - 9 738 64
XM_017020614.1 XP_016876103.1 7 - 8 589 64
XM_017020615.1 XP_016876104.1 8 - 9 738 64
XM_017020616.1 XP_016876105.1 8 - 9 738 64
GenBank mRNA AF385926.1 - 8 - 9 675 64
AK225288.1 - 8 - 9 643 64
AK225299.1 - 8 - 9 64
AK296771.1 - 7 - 8 563 64
AK298318.1 - 8 - 9 665 64
AK315503.1 - 8 - 9 641 64
BC000140.2 - 8 - 9 64
X14608.1 - 8 - 9 610 64

View Full Product Details