The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286839.1 | NP_001273768.1 | 13 - 14 | 1654 | 84 |
NM_005766.3 | NP_005757.1 | 13 - 14 | 1833 | 84 | |
XM_011521046.2 | XP_011519348.1 | 13 - 14 | 1833 | 84 | |
XM_017020312.1 | XP_016875801.1 | 13 - 14 | 1577 | 84 | |
XM_017020313.1 | XP_016875802.1 | 12 - 13 | 2570 | 84 | |
GenBank mRNA | AB008430.1 | - | 13 - 14 | 1463 | 84 |
BC041595.1 | - | 13 - 14 | 1680 | 84 | |
BC071592.1 | - | 13 - 14 | 1654 | 84 |