The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286839.1 | NP_001273768.1 | 22 - 23 | 2768 | 134 |
NM_005766.3 | NP_005757.1 | 21 - 22 | 2854 | 134 | |
XM_011521046.2 | XP_011519348.1 | 22 - 23 | 2947 | 134 | |
XM_017020312.1 | XP_016875801.1 | 21 - 22 | 2598 | 134 | |
XM_017020313.1 | XP_016875802.1 | 21 - 22 | 3684 | 134 | |
GenBank mRNA | AB008430.1 | - | 21 - 22 | 2484 | 134 |
BC041595.1 | - | 21 - 22 | 2701 | 134 | |
BC071592.1 | - | 22 - 23 | 2768 | 134 |