Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FARP1
Entrez Gene ID
10160
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 1
Gene Aliases
CDEP, FARP1-IT1, PLEKHC2, PPP1R75
Chromosome Location
Chr.13: 98142562 - 98449773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286839.1 NP_001273768.1 6 - 7 733 63
NM_005766.3 NP_005757.1 6 - 7 912 63
XM_011521046.2 XP_011519348.1 6 - 7 912 63
XM_017020312.1 XP_016875801.1 6 - 7 656 63
XM_017020313.1 XP_016875802.1 5 - 6 1649 63
GenBank mRNA AB008430.1 - 6 - 7 542 63
BC041595.1 - 6 - 7 759 63
BC071592.1 - 6 - 7 733 63

View Full Product Details