Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
Chromosome Location
Chr.19: 58346806 - 58353499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130786.3 NP_570602.2 2 - 3 128 113
GenBank mRNA AB073611.1 - 2 - 3 121 113
AF414429.1 - 2 - 3 108 113
AK055885.1 - 2 - 3 121 113
AK289417.1 - 2 - 3 92 113
BX325198.2 - 2 - 3 128 113
EU794585.1 - 2 - 3 120 113

View Full Product Details