Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FES
Entrez Gene ID
2242
Gene Name
FES proto-oncogene, tyrosine kinase
Gene Aliases
FPS
Chromosome Location
Chr.15: 90884421 - 90895776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143783.1 NP_001137255.1 10 - 11 1478 88
NM_001143784.1 NP_001137256.1 10 - 11 1442 88
NM_001143785.1 NP_001137257.1 10 - 11 1387 88
NM_002005.3 NP_001996.1 12 - 13 1748 88
XM_005254880.1 XP_005254937.1 11 - 12 1563 88
XM_005254882.1 XP_005254939.1 11 - 12 1554 88
XM_017022005.1 XP_016877494.1 12 - 13 1732 88
XM_017022006.1 XP_016877495.1 11 - 12 1605 88
XM_017022007.1 XP_016877496.1 11 - 12 1522 88
XM_017022008.1 XP_016877497.1 10 - 11 1396 88
XM_017022009.1 XP_016877498.1 12 - 13 1829 88
XM_017022010.1 XP_016877499.1 11 - 12 1680 88
GenBank mRNA AK300595.1 - 10 - 11 1377 88
AK300605.1 - 12 - 13 1706 88
AK312545.1 - 12 - 13 1737 88
AY513654.1 - 9 - 10 1268 88
AY513655.1 - 9 - 10 1268 88
AY513656.1 - 10 - 11 1478 88
AY513657.1 - 10 - 11 1442 88
BC035357.1 - 12 - 13 1724 88
X52192.1 - 12 - 13 1727 88

View Full Product Details