The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143783.1 | NP_001137255.1 | 12 - 13 | 1652 | 78 |
NM_001143784.1 | NP_001137256.1 | 12 - 13 | 1616 | 78 | |
NM_001143785.1 | NP_001137257.1 | 12 - 13 | 1561 | 78 | |
NM_002005.3 | NP_001996.1 | 14 - 15 | 1922 | 78 | |
XM_005254880.1 | XP_005254937.1 | 13 - 14 | 1737 | 78 | |
XM_005254882.1 | XP_005254939.1 | 13 - 14 | 1728 | 78 | |
XM_017022005.1 | XP_016877494.1 | 14 - 15 | 1906 | 78 | |
XM_017022006.1 | XP_016877495.1 | 13 - 14 | 1779 | 78 | |
XM_017022007.1 | XP_016877496.1 | 13 - 14 | 1696 | 78 | |
XM_017022008.1 | XP_016877497.1 | 12 - 13 | 1570 | 78 | |
XM_017022009.1 | XP_016877498.1 | 14 - 15 | 2003 | 78 | |
XM_017022010.1 | XP_016877499.1 | 13 - 14 | 1854 | 78 | |
GenBank mRNA | AK300595.1 | - | 12 - 13 | 1551 | 78 |
AK300605.1 | - | 14 - 15 | 1880 | 78 | |
AK312545.1 | - | 14 - 15 | 1911 | 78 | |
AY513654.1 | - | 11 - 12 | 1442 | 78 | |
AY513656.1 | - | 12 - 13 | 1652 | 78 | |
AY513657.1 | - | 12 - 13 | 1616 | 78 | |
BC035357.1 | - | 14 - 15 | 1898 | 78 | |
X52192.1 | - | 14 - 15 | 1901 | 78 |