The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166018.1 | NP_001159490.1 | 5 - 6 | 659 | 64 |
NM_007252.3 | NP_009183.3 | 5 - 6 | 659 | 64 | |
XM_011515113.2 | XP_011513415.1 | 4 - 5 | 679 | 64 | |
GenBank mRNA | U91935.1 | - | 4 - 5 | 599 | 64 |