The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202475.1 | NP_001189404.1 | 7 - 8 | 851 | 102 |
NM_001202481.1 | NP_001189410.1 | 8 - 9 | 1078 | 102 | |
NM_001202482.1 | NP_001189411.1 | 6 - 7 | 942 | 102 | |
NM_001202483.1 | NP_001189412.1 | 6 - 7 | 945 | 102 | |
NM_001883.4 | NP_001874.2 | 6 - 7 | 945 | 102 | |
XM_011515127.2 | XP_011513429.1 | 6 - 7 | 752 | 102 | |
XM_011515128.2 | XP_011513430.1 | 6 - 7 | 752 | 102 | |
XM_011515129.2 | XP_011513431.1 | 6 - 7 | 752 | 102 | |
XM_017011751.1 | XP_016867240.1 | 6 - 7 | 749 | 102 | |
XM_017011752.1 | XP_016867241.1 | 6 - 7 | 1889 | 102 | |
GenBank mRNA | AF011406.1 | - | 7 - 8 | 851 | 102 |
AF019381.1 | - | 6 - 7 | 809 | 102 | |
AK313661.1 | - | 6 - 7 | 906 | 102 | |
AY449734.1 | - | 6 - 7 | 700 | 102 | |
BC096830.1 | - | 6 - 7 | 709 | 102 | |
EU012439.1 | - | 6 - 7 | 700 | 102 | |
EU012440.1 | - | 6 - 7 | 700 | 102 | |
EU012441.1 | - | 6 - 7 | 700 | 102 | |
EU012442.1 | - | 5 - 6 | 611 | 102 | |
EU012443.1 | - | 6 - 7 | 697 | 102 | |
FJ200484.1 | - | 5 - 6 | 611 | 102 | |
U34587.1 | - | 6 - 7 | 700 | 102 |