The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076552.2 | NP_001070020.2 | 10 - 11 | 1307 | 90 |
NM_001242393.1 | NP_001229322.1 | 9 - 10 | 1144 | 90 | |
NM_018677.3 | NP_061147.1 | 9 - 10 | 1268 | 90 | |
XM_005260455.1 | XP_005260512.1 | 10 - 11 | 1307 | 90 | |
XM_005260456.1 | XP_005260513.1 | 9 - 10 | 1268 | 90 | |
XM_006723826.1 | XP_006723889.1 | 10 - 11 | 1183 | 90 | |
XM_011528905.1 | XP_011527207.1 | 11 - 12 | 1299 | 90 | |
XM_011528906.1 | XP_011527208.1 | 11 - 12 | 1299 | 90 | |
XM_011528907.1 | XP_011527209.1 | 10 - 11 | 1260 | 90 | |
XM_011528908.1 | XP_011527210.1 | 10 - 11 | 1260 | 90 | |
XM_011528909.2 | XP_011527211.1 | 11 - 12 | 1185 | 90 | |
XM_011528911.1 | XP_011527213.1 | 10 - 11 | 1194 | 90 | |
XM_011528912.2 | XP_011527214.1 | 11 - 12 | 1191 | 90 | |
GenBank mRNA | AF263614.1 | - | 9 - 10 | 1147 | 90 |
AK000162.1 | - | 9 - 10 | 1083 | 90 | |
AK000188.1 | - | 9 - 10 | 1221 | 90 | |
AK022608.1 | - | 3 - 4 | 380 | 90 | |
AK025990.1 | - | 9 - 10 | 1203 | 90 | |
AK092281.1 | - | 10 - 11 | 1219 | 90 | |
AK098026.1 | - | 4 - 5 | 743 | 90 | |
AL359946.1 | - | 8 - 9 | 977 | 90 | |
BC010141.2 | - | 6 - 7 | 647 | 90 | |
BC012172.1 | - | 9 - 10 | 1191 | 90 | |
BC073846.1 | - | 9 - 10 | 1032 | 90 | |
BC098422.1 | - | 8 - 9 | 863 | 90 | |
CR749716.1 | - | 8 - 9 | 977 | 90 |