The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017714.2 | NP_060184.2 | 4 - 5 | 396 | 118 |
NR_136628.1 | 4 - 5 | 396 | 118 | ||
NR_136629.1 | 4 - 5 | 396 | 118 | ||
XM_017027929.1 | XP_016883418.1 | 4 - 5 | 392 | 118 | |
XM_017027930.1 | XP_016883419.1 | 4 - 5 | 390 | 118 | |
XM_017027931.1 | XP_016883420.1 | 4 - 5 | 392 | 118 | |
GenBank mRNA | AK000219.1 | - | 3 - 4 | 363 | 118 |
AK025671.1 | - | 3 - 4 | 363 | 118 | |
AL601028.1 | - | 4 - 5 | 400 | 118 | |
BC025266.1 | - | 4 - 5 | 376 | 118 |