Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TASP1
Entrez Gene ID
55617
Gene Name
taspase 1
Gene Aliases
C20orf13, dJ585I14.2
Chromosome Location
Chr.20: 13104760 - 13638936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017027928.1 XP_016883417.1 2 - 3 141 113

View Full Product Details