Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCAN1
Entrez Gene ID
1827
Gene Name
regulator of calcineurin 1
Gene Aliases
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1
Chromosome Location
Chr.21: 34516442 - 34615142 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203418.2 NP_981963.1 1 - 2 346 80
XM_005260930.1 XP_005260987.1 1 - 2 290 80
GenBank mRNA AK293293.1 - 1 - 2 160 80
AK299444.1 - 1 - 2 96 80
BX419260.2 - 1 - 2 299 80
CR542065.1 - 1 - 2 85 80
DC331441.1 - 1 - 2 346 80
U85267.2 - 1 - 2 228 80

View Full Product Details