The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278225.1 | NP_001265154.1 | 3 - 4 | 324 | 104 |
NM_017987.4 | NP_060457.4 | 4 - 5 | 825 | 104 | |
NR_103475.1 | 4 - 5 | 673 | 104 | ||
NR_103476.1 | 4 - 5 | 498 | 104 | ||
XM_005269955.4 | XP_005270012.1 | 3 - 4 | 404 | 104 | |
XM_005269956.4 | XP_005270013.1 | 4 - 5 | 825 | 104 | |
XM_006717911.3 | XP_006717974.1 | 4 - 5 | 496 | 104 | |
XM_011539942.2 | XP_011538244.1 | 4 - 5 | 1237 | 104 | |
GenBank mRNA | AB040970.1 | - | 4 - 5 | 434 | 104 |
AF411980.1 | - | 3 - 4 | 345 | 104 | |
AF461266.1 | - | 3 - 4 | 404 | 104 | |
AK127019.1 | - | 4 - 5 | 825 | 104 | |
AK294211.1 | - | 3 - 4 | 307 | 104 | |
AY528716.1 | - | 4 - 5 | 463 | 104 | |
BC062660.1 | - | 4 - 5 | 420 | 104 | |
DA858392.1 | - | 4 - 5 | 418 | 104 |