The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007231.2 | NP_001007232.2 | 8 - 9 | 1423 | 82 |
NM_001166276.1 | NP_001159748.1 | 7 - 8 | 1363 | 82 | |
NM_001166277.1 | NP_001159749.1 | 7 - 8 | 1303 | 82 | |
NM_014882.2 | NP_055697.1 | 7 - 8 | 1360 | 82 | |
XM_005264675.2 | XP_005264732.1 | 8 - 9 | 1127 | 82 | |
XM_005264676.2 | XP_005264733.1 | 7 - 8 | 1011 | 82 | |
XM_011533206.2 | XP_011531508.1 | 8 - 9 | 1171 | 82 | |
XM_011533207.2 | XP_011531509.1 | 8 - 9 | 1137 | 82 | |
XM_011533208.2 | XP_011531510.1 | 9 - 10 | 1188 | 82 | |
XM_011533209.2 | XP_011531511.1 | 5 - 6 | 728 | 82 | |
XM_011533210.2 | XP_011531512.1 | 3 - 4 | 255 | 82 | |
XM_017005423.1 | XP_016860912.1 | 8 - 9 | 1031 | 82 | |
XM_017005424.1 | XP_016860913.1 | 8 - 9 | 1028 | 82 | |
XM_017005425.1 | XP_016860914.1 | 7 - 8 | 914 | 82 | |
XM_017005426.1 | XP_016860915.1 | 5 - 6 | 1365 | 82 | |
XM_017005427.1 | XP_016860916.1 | 5 - 6 | 1362 | 82 | |
GenBank mRNA | AK290396.1 | - | 8 - 9 | 1101 | 82 |
AK297056.1 | - | 7 - 8 | 1181 | 82 | |
AK297157.1 | - | 8 - 9 | 1163 | 82 | |
AK298669.1 | - | 7 - 8 | 1190 | 82 | |
BC039591.1 | - | 7 - 8 | 1224 | 82 | |
D29642.1 | - | 7 - 8 | 1173 | 82 | |
EU794656.1 | - | 7 - 8 | 1360 | 82 |