The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015043.3 | NP_055858.2 | 10 - 11 | 1822 | 59 |
NM_198868.2 | NP_942568.2 | 10 - 11 | 1822 | 59 | |
GenBank mRNA | AB014576.1 | - | 10 - 11 | 1822 | 59 |
AB449893.1 | - | 10 - 11 | 1785 | 59 | |
BC110400.1 | - | 1 - 2 | 42 | 59 | |
BC129994.1 | - | 10 - 11 | 1826 | 59 |