Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D9B
Entrez Gene ID
23061
Gene Name
TBC1 domain family member 9B
Gene Aliases
GRAMD9B
Chromosome Location
Chr.5: 179862071 - 179907856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015043.3 NP_055858.2 2 - 3 270 73
NM_198868.2 NP_942568.2 2 - 3 270 73
GenBank mRNA AB014576.1 - 2 - 3 270 73
AB449893.1 - 2 - 3 233 73
BC129994.1 - 2 - 3 274 73
BM459848.1 - - 285 73
BU183640.1 - 2 - 3 283 73

View Full Product Details