The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141973.2 | NP_001135445.1 | 5 - 6 | 701 | 62 |
NM_001141974.2 | NP_001135446.1 | 5 - 6 | 701 | 62 | |
XM_005245811.1 | XP_005245868.1 | 5 - 6 | 701 | 62 | |
XM_006710513.1 | XP_006710576.1 | 5 - 6 | 701 | 62 | |
XM_017000846.1 | XP_016856335.1 | 5 - 6 | 701 | 62 | |
XM_017000849.1 | XP_016856338.1 | 5 - 6 | 701 | 62 | |
GenBank mRNA | AK290210.1 | - | 5 - 6 | 657 | 62 |
AY461712.1 | - | 5 - 6 | 515 | 62 | |
BC030267.1 | - | 5 - 6 | 644 | 62 |