The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012325.2 | NP_036457.1 | 2 - 3 | 260 | 82 |
XM_011528696.2 | XP_011526998.1 | 2 - 3 | 262 | 82 | |
GenBank mRNA | AK312590.1 | - | 2 - 3 | 177 | 82 |
BC040410.1 | - | 2 - 3 | 187 | 82 | |
BC106068.1 | - | 2 - 3 | 190 | 82 | |
BC109281.3 | - | 2 - 3 | 184 | 82 | |
BC128442.1 | - | 2 - 3 | 155 | 82 | |
BJ992159.1 | - | 2 - 3 | 260 | 82 | |
U24166.1 | - | 2 - 3 | 185 | 82 |