Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YPEL5
Entrez Gene ID
51646
Gene Name
yippee like 5
Gene Aliases
CGI-127
Chromosome Location
Chr.2: 30146568 - 30160533 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127399.1 NP_001120871.1 3 - 4 407 73
NM_001127400.1 NP_001120872.1 3 - 4 643 73
NM_001127401.1 NP_001120873.1 4 - 5 704 73
NM_016061.2 NP_057145.1 2 - 3 346 73
XM_017004318.1 XP_016859807.1 4 - 5 942 73
XM_017004319.1 XP_016859808.1 3 - 4 380 73
XM_017004320.1 XP_016859809.1 3 - 4 881 73
XM_017004321.1 XP_016859810.1 3 - 4 573 73
XM_017004322.1 XP_016859811.1 3 - 4 804 73
GenBank mRNA AB098739.1 - 1 - 2 143 73
AF135161.1 - 2 - 3 250 73
AF151885.1 - 2 - 3 268 73
AF172940.1 - 2 - 3 270 73
AK025730.1 - 3 - 4 284 73
BC000836.1 - 3 - 4 221 73
BC047237.1 - 3 - 4 804 73

View Full Product Details