The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098635.1 | NP_001092105.1 | 15 - 16 | 3044 | 112 |
NM_001290202.1 | NP_001277131.1 | 15 - 16 | 2880 | 112 | |
NM_178860.4 | NP_849191.3 | 15 - 16 | 3044 | 112 | |
GenBank mRNA | AF502129.1 | - | 15 - 16 | 3132 | 112 |
AK054913.1 | - | 6 - 7 | 905 | 112 | |
AK055383.1 | - | 9 - 10 | 1316 | 112 | |
AK091522.1 | - | 15 - 16 | 2880 | 112 | |
AK096473.1 | - | 14 - 15 | 2293 | 112 | |
AK125377.1 | - | 15 - 16 | 3005 | 112 | |
AK223620.1 | - | 15 - 16 | 3006 | 112 | |
AL834405.1 | - | 14 - 15 | 2789 | 112 | |
AY038048.1 | - | 15 - 16 | 3035 | 112 |