The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286242.1 | NP_001273171.1 | 11 - 12 | 1946 | 74 |
NM_001520.3 | NP_001511.2 | 11 - 12 | 1946 | 74 | |
XM_017023188.1 | XP_016878677.1 | 11 - 12 | 1942 | 74 | |
GenBank mRNA | BC137229.1 | - | 11 - 12 | 1925 | 74 |
BX648143.1 | - | 11 - 12 | 1916 | 74 | |
CR627430.1 | - | 11 - 12 | 1930 | 74 | |
U02619.1 | - | 11 - 12 | 1966 | 74 | |
U06485.1 | - | 10 - 11 | 1703 | 74 |