The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286242.1 | NP_001273171.1 | 21 - 22 | 3470 | 77 |
NM_001520.3 | NP_001511.2 | 21 - 22 | 3470 | 77 | |
XM_017023188.1 | XP_016878677.1 | 21 - 22 | 3466 | 77 | |
GenBank mRNA | BC044857.2 | - | 1 - 2 | 112 | 77 |
BC137229.1 | - | 21 - 22 | 3449 | 77 | |
BX648143.1 | - | 21 - 22 | 3439 | 77 | |
CR627430.1 | - | 21 - 22 | 3455 | 77 | |
U02619.1 | - | 21 - 22 | 3490 | 77 |