The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006910.4 | NP_008841.2 | 15 - 16 | 2983 | 141 |
GenBank mRNA | AB112074.1 | - | 15 - 16 | 1987 | 141 |
AF352051.1 | - | 10 - 11 | 1506 | 141 | |
AK303040.1 | - | 13 - 14 | 1881 | 141 | |
BX648661.1 | - | 15 - 16 | 1923 | 141 | |
X85133.1 | - | 10 - 11 | 1506 | 141 |