The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053034.2 | NP_444262.1 | 13 - 14 | 1400 | 151 |
GenBank mRNA | AF421380.1 | - | 13 - 14 | 1147 | 151 |