The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176822.3 | NP_789792.1 | 4 - 5 | 2274 | 93 |
XM_011520044.1 | XP_011518346.1 | 4 - 5 | 2297 | 93 | |
GenBank mRNA | AK310900.1 | - | 4 - 5 | 2274 | 93 |
AY154469.1 | - | 3 - 4 | 1951 | 93 |