Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICAL2
Entrez Gene ID
9645
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases
MICAL-2, MICAL2PV1, MICAL2PV2
Chromosome Location
Chr.11: 12110576 - 12281017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282663.1 NP_001269592.1 15 - 16 2289 69
NM_001282664.1 NP_001269593.1 14 - 15 2142 69
NM_001282665.1 NP_001269594.1 14 - 15 2142 69
NM_001282666.1 NP_001269595.1 13 - 14 2067 69
NM_001282667.1 NP_001269596.1 13 - 14 2067 69
NM_014632.3 NP_055447.1 15 - 16 2293 69
XM_011520490.2 XP_011518792.1 13 - 14 1996 69
XM_011520491.2 XP_011518793.1 13 - 14 1996 69
XM_017018589.1 XP_016874078.1 14 - 15 2109 69
XM_017018590.1 XP_016874079.1 14 - 15 2154 69
XM_017018591.1 XP_016874080.1 14 - 15 2109 69
GenBank mRNA AB018293.1 - 15 - 16 2254 69
AB110785.1 - 13 - 14 1990 69
AB110786.1 - 13 - 14 1990 69
AK294845.1 - 14 - 15 2109 69
AK302580.1 - 6 - 7 751 69
AK302893.1 - 15 - 16 2371 69
BC044577.1 - 15 - 16 2278 69
BX538021.1 - 11 - 12 1817 69

View Full Product Details